Merck
  • Partial replacement of fishmeal and fish oil by algal meals in diets of red drum Sciaenops ocellatus
Partial replacement of fishmeal and fish oil by algal meals in diets of red drum Sciaenops ocellatus
Aquaculture (Amsterdam, Netherlands) (2018)
Perez-Velazquez M, et al.