Merck
[D-Ala2]deltorphin II analogs with high affinity and selectivity for delta-opioid receptor.
Biochemical and biophysical research communications (1991-10-31)
Y Sasaki, A Ambo, K Suzuki
ZUSAMMENFASSUNG
Nine [D-Ala2]deltorphin II (DL-II:Tyr-D-Ala-Phe-Glu-Val-Val-Gly-NH2) analogs having various aliphatic amino acids at positions 5 and 6 were synthesized to gain more information about the role of hydrophobic Val5,6 residues for the delta-opioid receptor selectivity. Binding assays of analogs replaced by Ala demonstrated the importance of hydrophobic Val5,6 residues in DL-II for delta-affinity and selectivity, and especially critical importance of Val5 residue for higher delta-selectivity. By enhancing the hydrophobicity of residues at positions 5 and 6, we have developed analogs with very high delta-affinity and selectivity over those of DL-II, e.g., [Ile5,6], [norleucine5,6] and [gamma-methyl-leucine5,6]DL-II, which will be useful as delta-selective ligands for investigation of the physiological role of opioid receptors.