Merck
  • The MDM2 RING and central acidic domains play distinct roles in MDM2 homodimerization and MDM2-MDMX heterodimerization
The MDM2 RING and central acidic domains play distinct roles in MDM2 homodimerization and MDM2-MDMX heterodimerization
The Journal of biological chemistry (2015)
Leslie PL, et al