Merck
  • Contrast-enhanced CT using a cationic contrast agent enables non-destructive assessment of the biochemical and biomechanical properties of mouse tibial plateau cartilage
Contrast-enhanced CT using a cationic contrast agent enables non-destructive assessment of the biochemical and biomechanical properties of mouse tibial plateau cartilage
Journal of Orthopaedic Research : Official Publication of the Orthopaedic Research Society (2016)
Lakin BA, et al.