Merck
  • Clenbuterol induces cardiac myocyte hypertrophy via paracrine signalling and fibroblast-derived IGF-1
Clenbuterol induces cardiac myocyte hypertrophy via paracrine signalling and fibroblast-derived IGF-1
Journal of Cardiovascular Translational Research (2010)
Bhavsar PK, et al.