Merck
  • Proper development of the outer longitudinal smooth muscle of the mouse pylorus requires Nkx2-5 and Gata3
Proper development of the outer longitudinal smooth muscle of the mouse pylorus requires Nkx2-5 and Gata3
Gastroenterology (2014)
Udager AM, et al.