- Relief of endoplasmic reticulum stress enhances DNA damage repair and improves development of pre-implantation embryos
Relief of endoplasmic reticulum stress enhances DNA damage repair and improves development of pre-implantation embryos
PLoS ONE (2017)
Dicks N, et al.