Merck
  • Examination of the Interaction between a Membrane Active Peptide
    and Artificial Bilayers by Dual Polarisation Interferometry
Examination of the Interaction between a Membrane Active Peptide<BR/> and Artificial Bilayers by Dual Polarisation Interferometry
Bio-protocol (2017)
Payne JAE, et al.