Merck
  • Potential role for the VDR agonist elocalcitol in metabolic control: Evidences in human skeletal muscle cells.
Potential role for the VDR agonist elocalcitol in metabolic control: Evidences in human skeletal muscle cells.
The Journal of steroid biochemistry and molecular biology (2017-01-04)
Cristina Antinozzi, Clarissa Corinaldesi, Carla Giordano, Annalinda Pisano, Bruna Cerbelli, Silvia Migliaccio, Luigi Di Luigi, Katia Stefanantoni, Gabriella Barbara Vannelli, Salvatore Minisola, Guido Valesini, Valeria Riccieri, Andrea Lenzi, Clara Crescioli
ZUSAMMENFASSUNG
Vitamin D plays a pivotal role to maintain skeletal muscle integrity and health. Vitamin D deficiency characterizes inflammatory myopathy (IM) and diabetes, often overlapping diseases involving skeletal muscle damage. Vitamin D receptor (VDR) agonists likely exert beneficial effects in both IM and metabolic disturbances. We aim to evaluate in vitro the effect of elocalcitol, a non-hypercalcemic VDR agonist, on the biomolecular metabolic machinery of human skeletal muscle cells (Hfsmc), vs. insulin (I). We analyzed GLUT4, Flotillin-1, Caveolin-3 and Caveolin-1 cell expression/localization; mTOR, AKT, ERK and 4E-BP1 phosphorylation; IL-6 myokine release; VDR expression. We investigated in vivo vitamin D status in IM subjects, evaluating VDR muscular expression and serum vitamin D with metabolism-related parameters, as glycemia, triglycerides, cholesterol, resistin and adiponectin. In Hfsmc, elocalcitol exerted an I-like effect, promoting GLUT4 re-localization in Flotillin-1, Caveolin-3 and Caveolin-1 positive sites and mTOR, AKT, ERK, 4E-BP1 activation; it enhanced IL-6 myokine release. IM subjects, all normoglycemic, showed VDR/vitamin D deficiency that, together with high lipidemic and resistin profile, possibly increases the risk to develop metabolic diseases. VDR agonists as elocalcitol may be therapeutic tools for skeletal muscle integrity/function maintenance, an indispensable condition for health homeostasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Elocalcitol, ≥98% (HPLC)