Merck
  • Partial biochemical characterization of Cu, Zn-superoxide dismutase extracted from eggs of hens (Gallus gallus domesticus)
Partial biochemical characterization of Cu, Zn-superoxide dismutase extracted from eggs of hens (Gallus gallus domesticus)
Food Chemistry (2017)
Wawrzykowski J and Kankofer M