Merck
  • Pupil dilation with intracameral epinephrine hydrochloride during phacoemulsification and intraocular lens implantation
Pupil dilation with intracameral epinephrine hydrochloride during phacoemulsification and intraocular lens implantation
Journal of Ophthalmology (2016)
Yu A, et al.