Merck
  • Sympathetic predominance is associated with impaired endothelial progenitor cells and tunneling nanotubes in controlled-hypertensive patients
Sympathetic predominance is associated with impaired endothelial progenitor cells and tunneling nanotubes in controlled-hypertensive patients
American Journal of Physiology. Heart and Circulatory Physiology (2014)
de Cavanagh EAMV, et al.