Merck
  • Polymorphism of Beta-Lactoglobulin Coding and 5?-Flanking Regions and Association with Milk Production Traits
Polymorphism of Beta-Lactoglobulin Coding and 5?-Flanking Regions and Association with Milk Production Traits
Biotechnology, Biotechnological Equipment (2012)
Zakizadeh S, et al.