Merck
  • Effect of dynamic high pressure on whey protein aggregation: A comparison with the effect of continuous short-time thermal treatments
Effect of dynamic high pressure on whey protein aggregation: A comparison with the effect of continuous short-time thermal treatments
Food Hydrocolloids (2008)
Gracia-Julia A, et al.