Merck
  • Application of fragment-based lead generation to the discovery of novel, cyclic amidine beta-secretase inhibitors with nanomolar potency, cellular activity, and high ligand efficiency.
Application of fragment-based lead generation to the discovery of novel, cyclic amidine beta-secretase inhibitors with nanomolar potency, cellular activity, and high ligand efficiency.
Journal of medicinal chemistry (2007-11-08)
Philip D Edwards, Jeffrey S Albert, Mark Sylvester, David Aharony, Donald Andisik, Owen Callaghan, James B Campbell, Robin A Carr, Gianni Chessari, Miles Congreve, Martyn Frederickson, Rutger H A Folmer, Stefan Geschwindner, Gerard Koether, Karin Kolmodin, Jennifer Krumrine, Russell C Mauger, Christopher W Murray, Lise-Lotte Olsson, Sahil Patel, Nate Spear, Gaochao Tian
ZUSAMMENFASSUNG
Fragment-based lead generation has led to the discovery of a novel series of cyclic amidine-based inhibitors of beta-secretase (BACE-1). Initial fragment hits with an isocytosine core having millimolar potency were identified via NMR affinity screening. Structure-guided evolution of these fragments using X-ray crystallography together with potency determination using surface plasmon resonance and functional enzyme inhibition assays afforded micromolar inhibitors. Similarity searching around the isocytosine core led to the identification of a related series of inhibitors, the dihydroisocytosines. By leveraging the knowledge of the ligand-BACE-1 recognition features generated from the isocytosines, the dihydroisocytosines were efficiently optimized to submicromolar potency. Compound 29, with an IC50 of 80 nM, a ligand efficiency of 0.37, and cellular activity of 470 nM, emerged as the lead structure for future optimization.