Merck
  • An insertion/deletion polymorphism within the 3'-untranslated region of COL1A2 confers susceptibility to osteoporosis
An insertion/deletion polymorphism within the 3'-untranslated region of COL1A2 confers susceptibility to osteoporosis
Molecular Medicine Reports (2016)
Jiang ZS and Hao ZH