Merck
  • A Modified Method for Detection of Post-Phoretic Activities in Mannose-(Mpi, EC 5.3. 1.8) and Glucose-(Gpi, EC 5.3. 1.9) 6-Phosphate Isomerases
A Modified Method for Detection of Post-Phoretic Activities in Mannose-(Mpi, EC 5.3. 1.8) and Glucose-(Gpi, EC 5.3. 1.9) 6-Phosphate Isomerases
Russian Journal of Genetics (2001)
Zasypkin MY, et al.