Merck
  • In-house phase determination of the lima bean trypsin inhibitor: a low-resolution sulfur-SAD case
In-house phase determination of the lima bean trypsin inhibitor: a low-resolution sulfur-SAD case
Acta crystallographica. Section D, Structural biology (2003)
Debreczeni J, et al.