Merck
  • Are cardiovascular and sympathoadrenal effects of human ?new pressor protein? preparations attributable to human coagulation beta-FXIIa?
Are cardiovascular and sympathoadrenal effects of human ?new pressor protein? preparations attributable to human coagulation beta-FXIIa?
American Journal of Physiology. Heart and Circulatory Physiology (2004)
Papageorgiou PC, et al.