Merck
  • Partial characterization of trypsin-like protease and molecular cloning of a trypsin-like precursor cDNA in salivary glands of Lygus lineolaris
Partial characterization of trypsin-like protease and molecular cloning of a trypsin-like precursor cDNA in salivary glands of Lygus lineolaris
Comparative Biochemistry and Physiology. Part B, Biochemistry & Molecular Biology (2002)
Zeng F, et al.