Merck
  • Proteomic profiles reveal age-related changes in coelomic fluid of sea urchin species with different life spans
Proteomic profiles reveal age-related changes in coelomic fluid of sea urchin species with different life spans
Experimental Gerontology (2013)
Bodnar A