Merck
  • Antinociceptive effect of intracerebroventricular administration of d-serine on formalin-induced pain
Antinociceptive effect of intracerebroventricular administration of d-serine on formalin-induced pain
Journal of Anesthesia (2014)
Ito M, et al.