Merck
Desmosomal proteins: new markers for identification and classification of tumors.
Laboratory investigation; a journal of technical methods and pathology (1986-01-01)
R Moll, P Cowin, H P Kapprell, W W Franke