Merck
  • Overexpression of the epidermal growth factor receptor confers migratory properties to nonmigratory postnatal neural progenitors.
Overexpression of the epidermal growth factor receptor confers migratory properties to nonmigratory postnatal neural progenitors.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2005-12-02)
Adan Aguirre, Tilat A Rizvi, Nancy Ratner, Vittorio Gallo
ZUSAMMENFASSUNG
Approaches to successful cell transplantation therapies for the injured brain involve selecting the appropriate neural progenitor type and optimizing the efficiency of the cell engraftment. Here we show that epidermal growth factor receptor (EGFR) expression enhances postnatal neural progenitor migration in vitro and in vivo. Migratory NG2-expressing (NG2+) progenitor cells of the postnatal subventricular zone (SVZ) express higher EGFR levels than nonmigratory, cortical NG2+ cells. The higher endogenous EGFR expression in SVZ NG2+ cells is causally related with their migratory potential in vitro as well as in vivo after cell engraftment. EGFR overexpression in cortical NG2+ cells by transient transfection converted these cells to a migratory phenotype in vitro and in vivo. Finally, cortical NG2+ cells purified from a transgenic mouse in which the EGFR is overexpressed under the CNP promoter exhibited enhanced migratory capability. These findings reveal a new role for EGFR in the postnatal brain and open new avenues to optimize cell engraftment for brain repair.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-EGFR-Antikörper, neutralisierend, Klon LA1, clone LA1, Upstate®, from mouse