Merck
  • Insulin acts through FOXO3a to activate transcription of plasminogen activator inhibitor type 1.
Insulin acts through FOXO3a to activate transcription of plasminogen activator inhibitor type 1.
Molecular Endocrinology
Ushma R Jag, Jiri Zavadil, Frederick M Stanley