- Cationic palladium complex catalyzed diastereo- and enantioselective tandem annulation of 2-formylarylboronic acids with allenoates.
Cationic palladium complex catalyzed diastereo- and enantioselective tandem annulation of 2-formylarylboronic acids with allenoates.
Organic letters (2009-09-02)
Xufen Yu, Xiyan Lu
PMID19719091
ZUSAMMENFASSUNG
Cationic palladium complex [(dppp)Pd(H(2)O)(2)](2+)(BF(4)(-))(2) catalyzed tandem annulation reactions of 2-formylarylboronic acids and allenoates produce the indenol derivatives diastereo- and enantioselectively in a highly efficient way.