Merck
  • Luminal leptin inhibits L-glutamine transport in rat small intestine: involvement of ASCT2 and B0AT1.
Luminal leptin inhibits L-glutamine transport in rat small intestine: involvement of ASCT2 and B0AT1.
American Journal of Physiology: Gastrointestinal and Liver Physiology
Ducroc R, Sakar Y, Fanjul C, Barber A, Bado A, Lostao MP