Merck
  • Differentiation of mesenchymal stem cells onto highly adherent radio frequency-sputtered carbonated hydroxylapatite thin films.
Differentiation of mesenchymal stem cells onto highly adherent radio frequency-sputtered carbonated hydroxylapatite thin films.
Journal of Biomedical Materials Research Part A (2010)
Sima, Livia E, et al.