Merck
  • SWI/SNF and Asf1 independently promote derepression of the DNA damage response genes under conditions of replication stress.
SWI/SNF and Asf1 independently promote derepression of the DNA damage response genes under conditions of replication stress.
Testing
Minard, LV; Lin, LJ; Schultz, MC
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Aktin-Antikörper, Klon C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Calmodulin-Bindungsprotein-Epitop-Tag-Antikörper, Upstate®, from rabbit