Merck
  • Defining the structural parameters that confer anticonvulsant activity by the site-by-site modification of (R)-N'-benzyl 2-amino-3-methylbutanamide.
Defining the structural parameters that confer anticonvulsant activity by the site-by-site modification of (R)-N'-benzyl 2-amino-3-methylbutanamide.
Journal of medicinal chemistry (2011-08-25)
Amber M King, Marc De Ryck, Rafal Kaminski, Anne Valade, James P Stables, Harold Kohn
ZUSAMMENFASSUNG
Primary amino acid derivatives (PAADs) (N'-benzyl 2-substituted 2-amino acetamides) are structurally related to functionalized amino acids (FAAs) (N'-benzyl 2-substituted 2-acetamido acetamides) but differ by the absence of the terminal N-acetyl group. Both classes exhibit potent anticonvulsant activities in the maximal electroshock seizure animal model, and the reported structure-activity relationships (SARs) of PAADs and FAAs differ in significant ways. Recently, we documented that PAAD efficacy was associated with a hydrocarbon moiety at the C(2)-carbon, while in the FAAs, a substituted heteroatom one atom removed from the C(2)-center was optimal. Previously in this issue, we showed that PAAD activity was dependent upon the electronic properties of the 4'-N'-benzylamide substituent, while FAA activity was insensitive to electronic changes at this site. In this study, we prepared analogues of (R)-N'-benzyl 2-amino-3-methylbutanamide to identify the structural components for maximal anticonvulsant activity. We demonstrated that the SAR of PAADs and FAAs diverged at the terminal amide site and that PAADs had considerably more structural latitude in the types of units that could be incorporated at this position, suggesting that these compounds function according to different mechanism(s).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Valproinsäure Natriumsalz, 98%
Sigma-Aldrich
5,5-Diphenylhydantoin, ≥98%