Merck
  • Inducing and characterizing liver regeneration in mice: Reliable models, essential "readouts" and critical perspectives.
Inducing and characterizing liver regeneration in mice: Reliable models, essential "readouts" and critical perspectives.
Current protocols in mouse biology
Mastellos, DC; Deangelis, RA; Lambris, JD
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-BrdU-Antikörper, Klon AH4H7-1 / 131-14871, Chemicon®, from mouse