Merck
  • Corneal wound healing and nerve morphology after excimer laser in situ keratomileusis in human eyes.
Corneal wound healing and nerve morphology after excimer laser in situ keratomileusis in human eyes.
Journal of Refractive Surgery (1996)
Latvala, T, et al.