- The phosphocholine-binding pocket on C-reactive protein is necessary for initial protection of mice against pneumococcal infection
The phosphocholine-binding pocket on C-reactive protein is necessary for initial protection of mice against pneumococcal infection
Test (2012)
Gang TB, et al.