Merck
  • Diosgenin induces cell cycle arrest and apoptosis in human leukemia K562 cells with the disruption of Ca 2+ homeostasis
Diosgenin induces cell cycle arrest and apoptosis in human leukemia K562 cells with the disruption of Ca 2+ homeostasis
Cancer Chemotherapy and Pharmacology (2005)
Liu MJ, et al.