Merck
  • Minimal methylated substrate and extended substrate range of Escherichia coli AlkB protein, a 1-methyladenine-DNA dioxygenase.
Minimal methylated substrate and extended substrate range of Escherichia coli AlkB protein, a 1-methyladenine-DNA dioxygenase.
The Journal of biological chemistry (2003-08-29)
Pertti Koivisto, Tod Duncan, Tomas Lindahl, Barbara Sedgwick
ZUSAMMENFASSUNG
The Escherichia coli AlkB protein, and two human homologs ABH2 and ABH3, directly demethylate 1-methyladenine and 3-methylcytosine in DNA. They couple Fe(II)-dependent oxidative demethylation of these damaged bases to decarboxylation of alpha-ketoglutarate. Here, we have determined the kinetic parameters for AlkB oxidation of 1-methyladenine in poly(dA), short oligodeoxyribonucleotides, nucleotides, and nucleoside triphosphates. Methylated poly(dA) was the preferred AlkB substrate of those tested. The oligonucleotide trimer d(Tp1meApT) and even 5'-phosphorylated 1-me-dAMP were relatively efficiently demethylated, and competed with methylated poly(dA) for AlkB activity. A polynucleotide structure was clearly not essential for AlkB to repair 1-methyladenine effectively, but a nucleotide 5' phosphate group was required. Consequently, 1-me-dAMP(5') was identified as the minimal effective AlkB substrate. The nucleoside triphosphate, 1-me-dATP, was inefficiently but actively demethylated by AlkB; a reaction with 1-me-ATP was even slower. E. coli DNA polymerase I Klenow fragment could employ 1-me-dATP as a precursor for DNA synthesis in vitro, suggesting that demethylation of alkylated deoxynucleoside triphosphates by AlkB could have biological significance. Although the human enzymes, ABH2 and ABH3, demethylated 1-methyladenine residues in poly(dA), they were inefficient with shorter substrates. Thus, ABH3 had very low activity on the trimer, d(Tp1meApT), whereas no activity was detected with ABH2. AlkB is known to repair methyl and ethyl adducts in DNA; to extend this substrate range, AlkB was shown to reduce the toxic effects of DNA damaging agents that generate hydroxyethyl, propyl, and hydroxypropyl adducts.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thrombin CleanCleave Kit