Merck
  • Nicotinamide phosphoribosyltransferase is essential for IL-1a-mediated dedifferentiation of articular chondrocytes via SIRT1 and ERK complex signaling
Nicotinamide phosphoribosyltransferase is essential for IL-1a-mediated dedifferentiation of articular chondrocytes via SIRT1 and ERK complex signaling
The Journal of Biological Chemistry (2011)
Hong,E et al.