Merck
  • Complement Factor H-ligand interactions: Self-association, multivalency and dissociation constants.
Complement Factor H-ligand interactions: Self-association, multivalency and dissociation constants.
Immunobiol. (2011)
Perkins SJ., et al.