Merck
Sleeping Beauty-mediated down-regulation of huntingtin expression by RNA interference.
Biochemical and biophysical research communications
Zongyu J Chen, Betsy T Kren, Phillip Y-P Wong, Walter C Low, Clifford J Steer