Merck
  • A novel protein kinase C alpha-dependent signal to ERK1/2 activated by alphaVbeta3 integrin in osteoclasts and in Chinese hamster ovary (CHO) cells.
A novel protein kinase C alpha-dependent signal to ERK1/2 activated by alphaVbeta3 integrin in osteoclasts and in Chinese hamster ovary (CHO) cells.
Journal of cell science (2005-07-15)
Nadia Rucci, Claudia DiGiacinto, Luigi Orrù, Danilo Millimaggi, Roland Baron, Anna Teti
ZUSAMMENFASSUNG
We identified a novel protein kinase C (PKC)alpha-dependent signal to extracellular signal-regulated kinase (ERK)1/2 in mouse osteoclasts and Chinese hamster ovary (CHO) cells, specifically activated by the alphaVbeta3 integrin. It involves translocation (i.e. activation) of PKCalpha from the cytosol to the membrane and/or the Triton X-100-insoluble subcellular fractions, with recruitment into a complex with alphaVbeta3 integrin, growth factor receptor-bound protein (Grb2), focal adhesion kinase (FAK) in CHO cells and proline-rich tyrosine kinase (PYK2) in osteoclasts. Engagement of alphavbeta3 integrin triggered ERK1/2 phosphorylation, but the underlying molecular mechanism was surprisingly independent of the well known Shc/Ras/Raf-1 cascade, and of phosphorylated MAP/ERK kinase (MEK)1/2, so far the only recognized direct activator of ERK1/2. In contrast, PKCalpha was involved in ERK1/2 activation because inhibition of its activity prevented ERK1/2 phosphorylation. The tyrosine kinase c-Src also contributed to ERK1/2 activation, however, it did not interact with PKCalpha in the same molecular complex. The alphaVbeta3/PKCalpha complex formation was fully dependent upon the intracellular calcium concentration ([Ca2+]i), and the use of the intracellular Ca2+ chelator 1,2-bis(o-amino-phenoxy)ethane-N,N,N',N'-tetraaceticacidtetra (acetoxymethyl) ester (BAPTA-AM) also inhibited PKCalpha translocation and ERK1/2 phosphorylation. Functional studies showed that alphaVbeta3 integrin-activated PKCalpha was involved in cell migration and osteoclast bone resorption, but had no effect on the ability of cells to attach to LM609, suggesting a role in events downstream of alphaVbeta3 integrin engagement.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
U0126, CAS-Nr. 109511-58-2, Calbiochem, U0126, CAS 109511-58-2, is a potent and specific inhibitor of MEK1 (IC50 = 72 nM) and MEK2 (IC50 = 58 nM). The inhibition is noncompetitive with respect to both ATP and ERK.
Millipore
BAPTA/AM, Membrane-permeable form of BAPTA.
Sigma-Aldrich
PP2, PP2, CAS 172889-27-9, is a potent, reversible, ATP-competitive, inhibitor of the Src family of protein tyrosine kinases (IC50 = 4, 5, 5, &100 nM for p56lck, p59fynT, Hck, & Src, respectively).
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
Gö 6976, Gö 6976, CAS 136194-77-9, is a cell-permeable, reversible, and ATP-competitive inhibitor of PKC (IC50 = 7.9 nM for rat brain). Exhibits selectively for PKCα (IC50 = 2.3 nM) and βI (IC50 = 6.2 nM).
Sigma-Aldrich
Ras-Aktivierungsassay-Kit
Sigma-Aldrich
Anti-GRB2-Antikörper, Klon 3F2, clone 3F2, Upstate®, from mouse