Merck
  • Molecular adjustment of the electronic properties of nanoporous electrodes in dye-sensitized solar cells.
Molecular adjustment of the electronic properties of nanoporous electrodes in dye-sensitized solar cells.
The journal of physical chemistry. B (2006-07-21)
Sven Rühle, Miri Greenshtein, S-G Chen, Alexandra Merson, Hillel Pizem, Chaim S Sukenik, David Cahen, Arie Zaban
ZUSAMMENFASSUNG
Molecular modification of dye-sensitized, mesoporous TiO2 electrodes changes their electronic properties. We show that the open-circuit voltage (V(oc)) of dye-sensitized solar cells varies linearly with the dipole moment of coadsorbed phosphonic, benzoic, and dicarboxylic acid derivatives. A similar dependence is observed for the short-circuit current density (I(sc)). Photovoltage spectroscopy measurements show a shift of the signal onset as a function of dipole moment. We explain the dipole dependence of the V(oc) in terms of a TiO2 conduction band shift with respect to the redox potential of the electrolyte, which is partially followed by the energy level of the dye. The I(sc) shift is explained by a dipole-dependent driving force for the electron current and a dipole-dependent recombination current.