- Ultrastructural and immunocharacterization of undifferentiated myocardial cells in the developing mouse heart.
Ultrastructural and immunocharacterization of undifferentiated myocardial cells in the developing mouse heart.
Journal of Cellular and Molecular Medicine
Zhang, F; Pasumarthi, KB
PMID17635645
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Atrial Natriuretic Peptide Antibody, clone A6, clone A6, Chemicon®, from mouse