Merck
  • Purification of 11 beta-hydroxysteroid dehydrogenase type 2 from human placenta utilizing a novel affinity labelling technique.
Purification of 11 beta-hydroxysteroid dehydrogenase type 2 from human placenta utilizing a novel affinity labelling technique.
The Biochemical journal (1996-02-01)
R W Brown, K E Chapman, P Murad, C R Edwards, J R Seckl
ZUSAMMENFASSUNG
11 beta-hydroxysteroid dehydrogenase type 2 (11 beta-HSD2) efficiently inactivates potent glucocorticoid hormones (cortisol and corticosterone), leaving aldosterone unmetabolized. Abundant 11 beta-HSD2 activity in human placenta plays a central role in controlling fetal glucocorticoid exposure, which if excessive is harmful and may predispose to low birth weight and hypertension in adulthood. Similar 11 beta-HSD2 activity in the distal nephron protects mineralocorticoid receptors from glucocorticoids and appears to be important in normal blood pressure control. We have purified human placental 11 beta-HSD2 16000-fold, to homogeneity, and determined over 100 residues of the internal amino acid sequence. Purification was assisted by a novel technique allowing highly specific (single spot on two-dimensional electrophoresis) photoaffinity labelling of active 11 beta-HSD2 in crude tissue extracts by its glucocorticoid substrates. This work reveals that 11 beta-HSD2 is a member of the short-chain alcohol dehydrogenase superfamily (apparent monomer M(r) approximately 40,000). It is a very basic (apparent pI = 9.1) intrinsic membrane protein, requiring as yet undefined membrane constituents for full stability. Affinity chromatography and affinity labelling studies suggest that 11 beta-HSD2 has a compulsory ordered mechanism, with NAD+ binding first, followed by a conformational change allowing glucocorticoid binding with high affinity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adenosin-5′-Monophosphat-Agarose, lyophilized powder