Merck
  • Homocysteine-induced endothelial cell adhesion is related to adenosine lowering and is not mediated by S-adenosylhomocysteine.
Homocysteine-induced endothelial cell adhesion is related to adenosine lowering and is not mediated by S-adenosylhomocysteine.
FEBS letters (2007-09-14)
Giovanna Cacciapuoti, Caterina Manna, Daniela Napoli, Vincenzo Zappia, Marina Porcelli
ZUSAMMENFASSUNG
Hyperhomocysteinemia is a cardiovascular risk factor and may contribute to the pathogenesis of atherosclerosis by altering endothelial functions. The mechanism of homocysteine-induced cell adhesion has been here investigated using EA.hy 926 cells. Homocysteine induces a stereospecific, time- and dose-dependent cell adhesion which is prevented by adenosine. The dramatic increase of S-adenosylhomocysteine induced by adenosine-2',3'-dialdehyde does not cause cell adhesion, indicating that no apparent relationship exists between this process and intracellular S-adenosylhomocysteine content. Homocysteine-induced cell adhesion is abolished by pre-treatment with adenosine-2',3'-dialdehyde, demonstrating that the adenosine depletion caused by reversal of S-adenosylhomocysteine hydrolase reaction is responsible for homocysteine-induced cell damage.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adenosine, periodate oxidized, ≥93%