Merck
  • Inosine 5′-monophosphate dehydrogenase activity as a biomarker in the field of transplantation.
Inosine 5′-monophosphate dehydrogenase activity as a biomarker in the field of transplantation.
Clinica Chimica Acta; International Journal of Clinical Chemistry (2011)
Glander P, Hambach P, et al.