Merck
  • Intracoronary delivery of umbilical cord blood derived unrestricted somatic stem cells is not suitable to improve LV function after myocardial infarction in swine.
Intracoronary delivery of umbilical cord blood derived unrestricted somatic stem cells is not suitable to improve LV function after myocardial infarction in swine.
Journal of Molecular and Cellular Cardiology
Amber D Moelker, Timo Baks, Kim M A M Wever, Dimitry Spitskovsky, Piotr A Wielopolski et al.