Merck
  • Upregulation of SOX9 in lung adenocarcinoma and its involvement in the regulation of cell growth and tumorigenicity.
Upregulation of SOX9 in lung adenocarcinoma and its involvement in the regulation of cell growth and tumorigenicity.
Clinical cancer research : an official journal of the American Association for Cancer Research
Jiang, SS; Fang, WT; Hou, YH; Huang, SF; Yen, BL; Chang, JL; Li, SM; Liu, HP; Liu et al.