- Subretinal delivery of recombinant AAV serotype 8 vector in dogs results in gene transfer to neurons in the brain.
Subretinal delivery of recombinant AAV serotype 8 vector in dogs results in gene transfer to neurons in the brain.
Molecular Therapy
Stieger, K; Colle, MA; Dubreil, L; Mendes-Madeira, A; Weber, M; Le Meur, G; Deschamps et al.
PMID18388922