Merck
  • Two-dimensional infrared correlation spectroscopic studies of polymer blends--specific interactions in polyethylene adipate/cholesteryl palmitate blends.
Two-dimensional infrared correlation spectroscopic studies of polymer blends--specific interactions in polyethylene adipate/cholesteryl palmitate blends.
Spectrochimica Acta Part A: Molecular Spectroscopy (2011)
Popescu MC, Vasile C.