- Optimizing annealing temperature overcomes bias in bisulfite PCR methylation analysis.
Optimizing annealing temperature overcomes bias in bisulfite PCR methylation analysis.
BioTechniques (2007-02-03)
Lanlan Shen, Yi Guo, Xinli Chen, Saira Ahmed, Jean-Pierre J Issa
PMID17269485