Merck
  • Proapoptotic effects of dietary (n-3) fatty acids are enhanced in colonocytes of manganese-dependent superoxide dismutase knockout mice.
Proapoptotic effects of dietary (n-3) fatty acids are enhanced in colonocytes of manganese-dependent superoxide dismutase knockout mice.
The Journal of Nutrition
Fan, YY; Zhan, Y; Aukema, HM; Davidson, LA; Zhou, L; Callaway, E; Tian, Y; Weeks et al.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Nitrotyrosin-Antikörper, Chemicon®, from rabbit